GC columns, Stabilwax-MS (fused silica)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
10674 10673
REST10674EA 814 EUR InStock
REST10674 REST10673
GC columns, Stabilwax-MS (fused silica)
Chromatographiesäulen
Stabilwax-MS columns ensure reproducible retention times from run to run, even with temperature cycling. When methods require trace analysis, this highly polar, low-bleed stationary phase produces excellent signal-to-noise levels. Ideal for food and flavor analysis (e.g., essential oils), fragrance and allergen analysis, as well as industrial solvent and chemical analysis.
  • High-polarity, stable polyethylene glycol (PEG) stationary phase
  • Low bleed and rugged enough to withstand repeated temperature cycles without retention time shifting
  • Temperature range: 40 to 250/260 °C
  • Equivalent to USP G14, G15, G16, G20 and G39 phases
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge