Columns, D3606 application set
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
83606-800 83606-800901 83606-810 83606-810901 83606-840 83606-850
REST83606-800EA 2150 EUR InStock
REST83606-800 REST83606-800901 REST83606-810 REST83606-810901 REST83606-840 REST83606-850
Columns, D3606 application set
Chromatographiesäulen
Packed, 2 column set gives complete resolution of benzene from ethanol, no compromising coelutions and for accurate quantification of benzene and toluene.
  • Individually tested for method applicability
  • ASTM method D3606 is alternative to TCEP columns
The column set is to accommodate both valve injection and/or syringe injection, inlet is identified on both column 1 and column 2.
Column 1:1,8 m length, 3,17 mm OD, 2,0 mm ID, nonpolar Rtx-1, configured with a 50,8 mm inlet void to facilitate on-column injection.
Column 2:4,9 m length, 3,17 mm OD, 2,0 mm ID, proprietary packing material, inlet of column is identified for proper orientation for connection to the valve.
Zertifizierungen: Conforms to the specifications established in the current ASTM method D3606 for the quantitation of benzene and toluene in spark ignition fuel containing ethanol.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge