Standard microliter syringes (Hamilton) for Agilent 7673, 7683, 7693A, and 6850 autosamplers
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
21221 21222 21223 20167 20168 20169 20170 20171 20172
REST21221EA 299 EUR InStock
REST21221 REST21222 REST21223 REST20167 REST20168 REST20169 REST20170 REST20171 REST20172
Standard microliter syringes (Hamilton) for Agilent 7673, 7683, 7693A, and 6850 autosamplers
Spritzen Spritzen für die Chromatographie
These Hamilton syringes are designed and tested to meet critical autosampler performance needs. Needle point styles are designed to withstand multiple, rapid injections through a septum.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge