Micro-vials with screw threads
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
21050 21051
REST21050EA 295 EUR InStock
REST21050 REST21051
Micro-vials with screw threads
Vials
These screw-thread micro-vials are come in a variety of sizes and volume capacities.
  • Borosilicate glass with graduated marking spot
  • Tapered for high recovery of contents
  • Open top cap attached
  • Used with Mininert® sampling valves
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge