Micro-volume syringes, positive displacement, (SGE)
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
24770 24771 24772 24773 24775 24776 24777 24778 24779
REST24770EA 208 EUR InStock
REST24770 REST24771 REST24772 REST24773 REST24775 REST24776 REST24777 REST24778 REST24779
Micro-volume syringes, positive displacement, (SGE)
Spritzen Spritzen für die Chromatographie
These micro-volume syringes have samples contained in the needle with full plunger displacement, ensuring no dead volume.
  • High precision and accuracy down to 0,1 µl
  • Seal between plunger and needle can be tightened for high-pressure injection
* 70 mm needle length, 0,47 mm OD, on-column syringe.
Bestellinformation: Replaceable needle/plunger kits and barrels are available on request.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge