Injector adapters
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
22318 22319 22320
REST22318EA 143 EUR InStock
REST22318 REST22319 REST22320
Injector adapters
Chromatographie-Zubehör Chromatographie-Ersatzteile
These injector adapters made of stainless steel are ideal for PerkinElmer CAP injector.
Siltek®-treated version available for increased inertness.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge