Adapters for capillary columns
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
24970 20544
REST24970EA 116 EUR InStock
REST24970 REST20544
Adapters for capillary columns
Chromatographie-Zubehör Chromatographie-Ersatzteile
These adapters are made of high quality stainless steel.
  • Ideal for Thermo Scientific TRACE and Focus SSL
  • Siltek®-treated version available for additional inertness
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge