Kettle, WK 3342
Lieferant: Severin
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
SEVEWK3342EA 176 EUR InStock
SEVEWK3342
Kettle, WK 3342
Heizgeräte Wasserkocher
This cordless, detachable jug kettle has a double walled stainless steel housing with hinged lid and easy pour spout for safe handling.
  • 4 temperatures settings: 80°, 85°, 90° and 100 °C
  • Heat insulated plastic handle
  • Auto switch-off
  • Transparent water level indicator on both sides
  • 360° connector for easy use and non-skid rubber feet
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge