Sample storage tubes, 2D barcoded
Lieferant: Thermo Scientific
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
MATRAB-1440EA 677 EUR InStock
MATRAB-1440
Sample storage tubes, 2D barcoded
2D barcoded storage tubes are designed for secure, individual sample identification. An encapsulated 2D barcode on each tube ensure safe, long term identification and tracking of samples, while 2D barcode readers allow simple decoding and integration into spreadsheets, databases and existing tracking systems.
  • 2D barcode is fully encapsulated behind an optically clear window to protect it against even the harshest conditions
  • V-shaped internal well design reduces dead volume and improves sample recovery
  • Rounded end caps and tapered rack positions ensure reliable 'pick and place' tube handling, idea for use with robotics
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge