PEEK tubing elbows
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
25308 25309
REST25308EA 32 EUR InStock
REST25308 REST25309
PEEK tubing elbows
Schläuche Chromatographie-Schläuche
Tubing elbows are ideal for routing 1,6 mm PEEK tubing that snap the tubing into the elbow. Its prevent pinching of PEEK tubing, which can cause high pressure.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge