DNA clean up, Sephadex® G-75 M
Lieferant: SIGMAALDRICH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
G75120-500G G75120-10G G75120-50G
SIALG75120-500GEA 2760 EUR InStock
SIALG75120-500G SIALG75120-10G SIALG75120-50G
DNA clean up, Sephadex® G-75 M
Nukleinsäure-Reagenzien Reinigungskits und Reagenzien für Nukleinsäure
Sephadex® G-75 is a gel filtration media used in gel filtration chromatography and protein chromatography. Sephadex® G-75 has been used to study cystatin of the mammalian brain, suggest candidates for development of novel oral or other anti-infective agents and to purify procoagulant metalloproteinase from rattlesnake venom.
  • Bead diameter of 40 to 120 µm
  • 1 g swells to 12 to 15 ml gel
  • Packed in poly bottle
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge