Pulse dampener, MiniPulse
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
25238 25239
REST25238EA 503 EUR InStock
REST25238 REST25239
Pulse dampener, MiniPulse
Chromatographie-Zubehör Chromatographie-Ersatzteile
The MiniPulse pulse damper is ideal for the applications where minimising the total system volume is critical.
  • Compact unit (64×38 mm) can be placed almost anywhere
  • Small, 160 µl dead volume at atmospheric pressure
  • PEEK unit can withstand pressures to 345 bar
  • 316 stainless steel unit can withstand pressures to 413 bar
Improves system baseline stability while increasing the total system volume by only 160 µl.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge