Chemical resistant overalls, MICROGARD® 2300 PLUS, model 132
Lieferant: ANSELL HEALTH CARE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
YY23T-00132-02 YY23T-00132-03 YY23T-00132-04 YY23T-00132-05 YY23T-00132-06 YY23T-00132-07
ANSEYY23T-00132-02EA 837 EUR InStock
ANSEYY23T-00132-02 ANSEYY23T-00132-03 ANSEYY23T-00132-04 ANSEYY23T-00132-05 ANSEYY23T-00132-06 ANSEYY23T-00132-07
Chemical resistant overalls, MICROGARD® 2300 PLUS, model 132
Einweg-Kleidung
These overalls are made from a PE barrier coating with a bi-component non woven inner layer, the combination of which provides an excellent barrier to many harmful chemicals while being lightweight and yet relatively strong and durable.
  • Elasticated 3-piece hood, wrists, waist and ankles
  • Two-way front zipper with resealable storm flap
  • Finger loops
  • Bright yellow colour for improved worker safety
  • Lightweight yet relatively strong and durable
  • Stitched and taped seams for increased strength and an effective barrier to liquids and particulates
  • Polycoated bi-component barrier resists permeation of numerous liquid chemicals
Typical applications include: Environmental clean-up, sewage purification installations, industrial and chemical manufacturing, industrial cleaning.
Zertifizierungen: Type 3-B/4-B/5-B, EN 14126, EN 1073-2, EN 1149-5.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge