Natural gas standards
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
34438 34439 34440
REST34438EA 0 EUR InStock
REST34438 REST34439 REST34440
Natural gas standards
Standards Umweltanalyse-Standards
Three mixes, from lean to rich.
Standard #1: Nitrogen, 1.000 %; carbon dioxide, 0.500 %; methane UHP, 94.750 %; ethane UHP, 2.000 %; propane, 0.750 %; isobutane, 0.300 %; n-utane, 0.300 %; isopentane, 0.150 %; n-pentane, 0.150 %; hexanes plus**, 0.100 %
Standard #2: Nitrogen, 2.500 %; carbon dioxide, 1.000 %; methane UHP, 85.250 %; ethane UHP, 5.000 %; propane, 3.000 %; isobutane, 1.000 %; n-butane, 1.000 %; isopentane, 0.500 %; n-pentane, 0.500 %; hexanes plus**, 0.250 %
Standard #3: Nitrogen, 5.500 %; carbon dioxide, 1.500 %; methane UHP, 70.000 %; ethane UHP, 9.000 %; propane, 6.000 %; isobutane, 3.000 %; n-butane, 3.000 %; isopentane, 1.000 %; n-pentane, 1.000 %; hexanes plus**, 0.500 %
Precise concentrations are provided on the data sheet included with each cylinder and may vary slightly from those listed here.
**A complete list of hexanes plus is available.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge