FSC 22® embedding compound, Surgipath®
Lieferant: Leica
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
Surgipath®
LEIC3801480EA 0 EUR InStock
LEIC3801480
FSC 22® embedding compound, Surgipath®
Färbemittel Mikroskopie- und Färbereagenzien
This FSC 22® is a water soluble embedding compound used in frozen sectioning. The compound bonds and encapsulates tissue specimens to the object holder for cryosectioning.
  • Provides consistent sectioning with minimal curling of sections
  • Working temperature −20 °C
  • Available in clear for better visualisation of small specimens
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge