Certified MS screw thread vial kits, 9 mm
Lieferant: THERMO NATIONAL SCIENTIFIC
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
MSCERT5000401W MSCERT5000S41W
NSCAMSCERT5000401WEA 380 EUR InStock
NSCAMSCERT5000401W NSCAMSCERT5000S41W
Certified MS screw thread vial kits, 9 mm
Vials
These certified clean, low particle, low background vials are suitable for high sensitivity LC/MS and GC/MS applications. They are made of 33 expansion borosilicate clear or amber glass (type 1, class A).
  • Tested and certified for 15 critical parameters
  • LC/MS and GC/MS chromatograms included in each pack
  • Pre-cleaned packaging protects product integrity
Reusable two compartment trays protect vials and closures while keeping matching supplies together.
Lieferumfang: Each kit includes 100 vials and 100 caps with pre-assembled septa and comes with a Certificate of Conformance.
Verpackung: These unassembled kits are packaged in clear trays.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge