Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Anti-ATXN8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Bioss

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

BOSSBS-11775R-FITCEA 437 EUR InStock
BOSSBS-11775R-FITC
Anti-ATXN8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Twinkle, also known as PEO1 (Progressive external ophthalmoplegia 1 protein), PEOA3, SANDO or TWINL, is a mitochondrial protein that functions as a 5’-3’ nucleotide-dependent DNA helicase. Colocalized with mtDNA (mitochondrial DNA) in mitochondrial nucleoids, Twinkle is important in the metabolism and maintenance of mtDNA, playing a crucial role in the regulation of mtDNA copy numbers. Twinkle is expressed at high levels in testis, pancreas and skeletal muscle and exists as three isoforms due to alternative splicing events. Defects in the gene encoding Twinkle are the cause of two conditions: progressive external ophthalmoplegia with mitochondrial DNA deletions autosomal dominant 3 (PEOA3) and sensory ataxic neuropathy dysarthria and ophthalmoparesis (SANDO). PEOA3 is characterized by ptosis and weak muscles, while SANDO is characterized by ophthalmoparesis, dysarthria and sensory ataxic neuropathies.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ATXN8
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now
 

PRODUKT

WEITERE INFORMATIONEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge