Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Fluid bed dryer, FBD 2000 digital

Lieferant: Endecotts

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

ENDED-FBDP2000/50EA 0 EUR InStock
ENDED-FBDP2000/50
Fluid bed dryer, FBD 2000 digital
The FBD 2000 fluid bed dryer is a compact, portable dryer with powerful air delivery system which makes drying a very fast operation.

  • Built in digital timer to pre-set the drying time
  • High airflow rates provide excellent heat transfer ensuring faster and more homogeneous drying
  • Precise controls ensure uniform and reproducible results
  • Suitable for most granular and powdered materials

The fluid bed dryer delivers the high volume air flow with a powerful fan from the base unit into a special tub assembly which holds the sample material. The flow of heated air passes through a diffuser gauze which supports the bed and evenly distributes the air as it passes into the tub. A filter bag at the top of the tub keeps the sample in while allowing the air, moisture and gases to escape. The fluidisation mixes and separates the particles, minimising the risk of abrasion and the creation of lumps resulting in a truly representative sample.

Temperature control : 0 to 200 °C
Drying time: 15 min

Zubehörinformationen: Single tubs come in 2 or 5 litre sizes in either stainless steel or glass. A multi-tub unit with 4×300 ml tubs is also available for drying four samples simultaneously. Glass tubs are particularly useful for observing the fluidisation process to establish optimum settings. Filter bags are usually nylon or terylene is available for more aggressive conditions such as sustained high temperature drying.

Order Now
 

PRODUKT

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge