Erucic acid methyl ester, Dr Ehrenstorfer
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
EHERC13203060EA 34 EUR InStock
EHERC13203060
Erucic acid methyl ester, Dr Ehrenstorfer
Standards Lebensmittel-Standards
These reference standards are unsaturated fatty acids and methyl esters in nature.
  • Liquid phase and colourless
Purity of these standards are determined by external standard method and purity was determined as 99,8%.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge