Disposable cutting blades for BioWelder®
Lieferant: Sartorius
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
SART16373EA 446 EUR InStock
SART16373
Disposable cutting blades for BioWelder®
Skalpelle Skalpellklingen
Single use stainless steel cutting blade to be used with BioWelder® (not BioWelder® TC).
  • With laser point 0,4 mm
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge