Surgical gown, Foliodress® Protect Reinforced
Lieferant: HARTMANN PAUL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
992817 992818 992819
HART992817EA 267 EUR InStock
HART992817 HART992818 HART992819
Surgical gown, Foliodress® Protect Reinforced
Einweg-Kleidung
These single-use surgical gowns made of a multilayer, fluid repellent, non woven fabric provide great resistance against the penetration of fluids and bacteria.
  • Half-sleeve reinforced surgical wrap-around gown with a reinforcement zone in the sleeves and chest area
  • Extra wide overlap at the back
  • Combitape® closing system to flexibly and securely adjust the neckline of the gown for an improved fit
  • Red neckline indicating high performance level for easier identification of the gown's protection level
  • With SMS-wrapping and paper towels
  • With ultra-sonic welded seams, heavy duty waist ties and elastic long knit cuffs
Ideal for operations involving large amounts of fluids.
Zertifizierungen: DIN EN 13795:2011+A1:2003
Verpackung: Individually packed.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge