Reference materials, pour point standards, VHG
Lieferant: Promochem
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
PROOVHG-P15-250EA 262 EUR InStock
PROOVHG-P15-250
Reference materials, pour point standards, VHG
These reference materials are intended for use in accordance with ASTM D97 only, for determination of the pour point of petroleum products.
Lieferumfang: Each standard is supplied with a full certificate of analysis (COA) that states the certified value.
Achtung! Bitte beachten: Not intended for use with phase instruments.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge