ORF Clones, GABA B receptor 2 (GABBR2), human tagged, TrueORF Gold
Lieferant: OriGene
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
ORIGRC219404EA 1690 EUR InStock
ORIGRC219404
ORF Clones, GABA B receptor 2 (GABBR2), human tagged, TrueORF Gold
Vektoren, Plasmide und Bibliotheken
GABA B receptor 2 (GABBR2), human tagged ORF clone was engineered to express the complete ORF with an expression tag. Expression varies depending on the nature of the gene.
The ORF clone is ion-exchange column purified, transfection-ready dried plasmid DNA.
he multi-pass membrane protein encoded by this gene belongs to the G-protein coupled receptor 3 family and GABA-B receptor subfamily. The GABA-B receptors inhibit neuronal activity through G protein-coupled second-messenger systems, which regulate the release of neurotransmitters, and the activity of ion channels and adenylyl cyclase. This receptor subunit forms an active heterodimeric complex with GABA-B receptor subunit 1, neither of which is effective on its own. Allelic variants of this gene have been associated with nicotine dependence.
Lieferumfang: Include 2 vector sequencing primers.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge