KORASILON™ pastes
Lieferant: Sigma-Aldrich
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
769711-1EA 769681-1EA
SIAL769711-1EAEA 46.3 EUR InStock
SIAL769711-1EA SIAL769681-1EA
KORASILON™ pastes
Schmiermittel
The pastes are excellently suitable as glide-, lubricant- and release agent for e. g. vales, joints and gaskets, ground joints, cables and wires. They are frequently also used as assembly aid as well as protection- and sealing aid. Furthermore, we recommend the pastes for maintaining rubber seals. In electronics, they find their use for the assembly of insulators and switchgears amongst others.
  • Stable at temperatures ranging from -50 °C to more than 300 °C
  • Exceptional hydrophobic and practically insoluble in water
  • Show thixotropic behavior
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge