ESD-Sicherheitsschuhe, Sandalen, Velcro®, Basic Comfort
Lieferant: Honeywell Safety Products
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
DALL98405-559/48EA 113 EUR InStock
DALL98405-559/48
ESD-Sicherheitsschuhe, Sandalen, Velcro®, Basic Comfort
ESD safety shoes made from water repellent nubuck and suede leather upper plus a durable, anti-slip, dual-density TPU/PU sole, resistant to oil and fuel.
  • Premium textile lining
  • Orthopaedic insole possible
  • Light density, shock-absorbent PU midsole
  • Protective steel toecap
Width: 9 (35-37); width: 11 (38-48)
Colour: Black/yellow
Zertifizierungen: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S1, SRC
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge