Scanning analysers
Lieferant: Palintest
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
705-0660EA 1620 EUR InStock
705-0660 705-0661
Scanning analysers
Analysatoren Elementar-Analysatoren
The SA1100 provides reliable monitoring for lead and copper in water using the voltametric method. Once connected to a PC, it can be used to access data stored in the instrument log, or to upgrade the instrument software.
  • Fixed voltage potentiostat
  • Rapid results reported in µg/l for copper or lead
  • Battery power saving system: Automatic switch-off after five minutes
Lieferumfang: Supplied with calibration chip, user instructions, and consumables pack for ten analyses in carrying case.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge