Caspase Substrate Set
Lieferant: PromoKine
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
POMOCA57711161000EA 599 EUR InStock
POMOCA57711161000 POMOCA57711031000 POMOPK-CA577-K138 POMOPK-CA707-10210 POMOPK-CA577-K137 POMOPK-CA577-K136 POMOCA57710691000 POMOCA57710701000 POMOPK-CA577-K135 POMOPK-CA577-K134 POMOCA57711101000 POMOPK-CA577-K133 POMOPK-CA577-K132 POMOPK-CA577-K131 POMOCA57711021000 POMOCA57710081000 POMOCA57710761000 POMOCA57710631000 POMOCA57710711000 POMOPK-CA707-10207 POMOCA57711131000 POMOCA57711141000 POMOCA57711011000 POMOPK-CA707-10202 POMOCA57711091000 POMOCA57710751000 POMOCA57711171000 POMOCA57711041000 POMOCA57710621000 POMOCA57710071000 POMOPK-CA577-K140 POMOCA57710721000
Caspase Substrate Set
The Caspase Substrate Sets can be used for setting up custom assays.
  • Ready-to-use form
  • Set up your own assays
Caspase substrates come in ready to use form for both fluorometric and colorimetric caspase assays. They can be used for setting up custom assays together with ready to use active caspases and caspase inhibitors.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge