Parallel synthesis kit, DrySyn®
Lieferant: Asynt
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
ASYNADS3-PS-17.4EA 0 EUR InStock
ASYNADS3-PS-17.4
Parallel synthesis kit, DrySyn®
The DrySyn parallel synthesis kit allows the scientist to heat and stir up to 27 small scale reactions in parallel using any standard hotplate.
  • Will fit Biotage 2 to 5 ml microwave tubes
  • No oil baths required for heating
  • Ideal for producing small, focused libraries of compounds
Lieferumfang: Kit Includes: Base plate to fit any hotplate stirrer, heat resistant handle and one pack of three standard reaction vial inserts.
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge