10130.14
10130.30
10130.A1
10130.14EA
28.8
EUR
InStock
10130.14
10130.30
10130.A1
Graphit 75 - 82% ≥75-82%, mikrokristallines Pulver -300 mesh 18-25% Asche
Graphit
CAS No.: 7782-42-5
Physical Form: Powder
Appearance: Black
Odour: Odourless
UN No.: 1325
MDL No.: MFCD00144065