Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Light duty orbital shakers, digital

Lieferant: OHAUS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

30391900 30391914
OHAU30391900EA 1180 EUR InStock
OHAU30391900 OHAU30391914
Light duty orbital shakers, digital
Schüttler Kreisschüttler
These light duty digital orbital shakers are ideal for a wide variety of shaking applications. Tray includes a non-skid rubber mat. The mat can be removed to mount a variety of optional flask clamps or test tube racks directly onto the tray.

  • Takes up less space and easily fits into most hoods and incubators
  • Variable speed microprocessor control provides consistent uniform shaking action and will display last set-point and will restart if power is interrupted
  • Speed ramping feature slowly increases speed to desired set-point to avoid splashing
  • Spill-resistant design ensures fluids are channelled away from internal components
  • Audible and visual signals will activate when system detects an obstruction or tray overload
  • In timed mode, alarm will sound when the time reaches zero
  • Can be used at −10 to 40/60 °C (max. 80% relative humidity, non condensing), in an incubator, CO₂ incubator or cold room

Touch pad controls with easy-to-read, independent LED displays for speed and time allow operator to view both settings at once. Provides repeatable and accurate results every time and is easily visible across laboratory benches. Timer will display elapsed time or, when programmed to user defined limit, will shut off unit when time reaches zero. Display will show last used settings, even after power has been turned off.

Triple eccentric drive: Permanently lubricated ball bearings and maintenance free, brushless DC motor provide reliable service and continuous duty operation.

Lieferumfang: Supplied with an 299×222 mm non-skid rubber mat and detachable, 3-wire cord and plug

Order Now
 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge