Recombinant SOLu-trypsin (from P. pastoris)
Lieferant: SIGMAALDRICH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
EMS0004-100UG EMS0004-4X100U
SIALEMS0004-100UGEA 93.7 EUR InStock
SIALEMS0004-100UG SIALEMS0004-4X100U
Recombinant SOLu-trypsin (from P. pastoris)
Enzyme
SOLu-Trypsin is an advanced proteomics grade enzyme that is solution stable for mass spectrometry. Recombinant, expressed in Pichia pastoris.
  • Ready to use - no preparation, such as reconstitution or thawing, is required
  • Fits seamlessly into established workflow - no need to modify protocols
  • Eliminates waste - remains stable in the refrigerator after use so there is no need to discard excess product
  • Recombinant, porcine sequence - no chymotryptic activity
  • Stable for short-term use at room temperature in an autosampler or on a liquid handling robot
SOLu-Trypsin is prepared from recombinant trypsin, porcine sequence. It is naturally devoid of chymotryptic activity. This high quality trypsin is suitable for proteomics use and is supplied in a ready to use solution.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge
Diese Webseite verwendet Cookies, eigene oder von Drittanbieter, zur Sammlung statistischer Informationen, um Inhalte und Anzeigen für Sie zu personalisieren. Wenn Sie diese Seite weiterhin verwenden, akzeptieren Sie die Cookie-Nutzung.
Weitere Informationen finden Sie in unseren Datenschutzbestimmungen..