Labels, laser Tough-Spots®
Lieferant: Sigma-Aldrich
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
L6164-1PAK L6289-1PAK L6414-1PAK L6539-1PAK L6664-1PAK L6789-1PAK L5414-1PAK L5539-1PAK L5664-1PAK L5789-1PAK L5914-1PAK L6039-1PAK T1316-1PAK
SIALL6164-1PAKEA 75.2 EUR InStock
SIALL6164-1PAK SIALL6289-1PAK SIALL6414-1PAK SIALL6539-1PAK SIALL6664-1PAK SIALL6789-1PAK SIALL5414-1PAK SIALL5539-1PAK SIALL5664-1PAK SIALL5789-1PAK SIALL5914-1PAK SIALL6039-1PAK SIALT1316-1PAK
Labels, laser Tough-Spots®
Etiketten
These Tough-Spots® polyolefin labels are specially top-coated to readily accept laser printing.
  • Temperature resistant from −196 to +150 °C
  • Chemically inert, resist most organic solvents and caustic agents
  • Adhere to most plastics, glass and metals without cracking, peeling or degrading
Tough-Spots® withstand freezer temperatures down to −196 °C (liquid and vapor phase nitrogen), boiling water baths (100 °C), autoclaving and moderate oven temperatures (150 °C).
Verpackung: 20 sheets per pack.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge