L09871.06
L09871.14
L09871.06EA
60
EUR
InStock
L09871.06
L09871.14
3-Bromphenylisothiocyanat ≥97%
3-Bromphenylisothiocyanat
Purity: 97%
CAS No.: 2131-59-1
Molecular Formula: C7H4BrNS
Formula Weight: 214.09
Flash Point: 110°(230°F)
Refractive Index: 1.6850
Odour: Pungent
Boiling Point: 255-256°
Beilstein Reference: 2081173
UN No.: 2922
MDL No.: MFCD00004803