Chemical resistant overalls, Body Tech T
Lieferant: BEST FRANCE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
01074T030XL 01074T030XXL 01074T030XXXL
BESF01074T030XLEA 0 EUR InStock
BESF01074T030XL BESF01074T030XXL BESF01074T030XXXL
Chemical resistant overalls, Body Tech T
Einweg-Kleidung
These hooded overalls made from laminated PE provide protection against light splashes and sprays of hazardous liquids as well as against hazardous dusts.
  • Elastic cuffs, ankles and back
  • Good moisture wicking properties
  • Zip closure (double slider) with adhesive flap
  • Waterproof and impervious to certain solvents or alcohols
  • Highly resistant seams with thermally bonded tape
  • Anti-static according to EN 1149-5
Weight: 58 g/m²
Zertifizierungen: Type 5/6, EN 1149-5; Cat. III
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge