A11999.06
A11999.14
A11999.06EA
48.6
EUR
InStock
A11999.06
A11999.14
3-Chlorphenylisothiocyanat ≥97%
3-Chlorphenylisothiocyanat
Purity: 97%
CAS No.: 2392-68-9
Molecular Formula: C7H4ClNS
Formula Weight: 169.64
Flash Point: 110°(230°F)
Refractive Index: 1.6585
Odour: Pungent
Boiling Point: 249-250°
Beilstein Reference: 636856
UN No.: 2922
MDL No.: MFCD00004805