B23900.0B
B23900.30
B23900.0BEA
170
EUR
InStock
B23900.0B
B23900.30
Isophthalonitril ≥98%
Isophthalonitril
Purity: 98%
CAS No.: 626-17-5
Molecular Formula: C8H4N2
Formula Weight: 128.13
Flash Point: 150°(302°F)
Odour: Almond-like
Boiling Point: 288°
Beilstein Reference: 1072209
MDL No.: MFCD00001795