Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

Nucleic acid purification system, maxXbond™

Lieferant: PanReac AppliChem

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: 0.0 von 5

Sortieren nach:

MB007.1000EA 343 EUR InStock
APLIMB007.1000
Nucleic acid purification system, maxXbond™
MaxXbond™ is 100% nucleic acid-free and is the first regeneration system for silica matrices for the isolation and purification of nucleic acid.

  • Biodegradable and non toxic
  • Full binding capacity after the regeneration
  • No damage to silica matrix
  • Savings of up to 70% when isolating nucleic acids

The silica columns used today are high tech products of high value. Not all nucleic acids are eluted from the silica matrix after the first usage is the reason that these valuable columns are not recycled. Usually, about 5 to 10% of the isolated nucleic acids remain trapped in the silica matrix. This residual DNA contains both free molecules and molecules complexed with protein particles or bacterial fragments. Therefore, even when reusing the columns for the preparation of the identical plasmid, DNA binding capacity is reduced. The used columns that are contaminated by recombinant DNA molecules and bacteria should be autoclaved to guarantee environmentally friendly disposal. As a result, laboratories that use DNA binding columns for the isolation and purification of nucleic acids not only dispose of a high-value item, but they must also deal with the costs of excessive biohazardous waste.

Order Now
 

PRODUKT

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge