Professional ventilated refrigerators, GKvesf
Lieferant: Liebherr
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
GKVESF5445 GKVESF4145
LIEBGKVESF5445EA 0 EUR InStock
LIEBGKVESF5445 LIEBGKVESF4145
Professional ventilated refrigerators, GKvesf
Kühlschränke
These commercial refrigerators feature a fan for forced air cooling with temperature adjustable from –2 to +15 °C or +1 to +15 °C.
  • Electronic temperature regulation, with digital display
  • Alarm for temperature and door opening
  • Automatic defrosting with evaporation of condensates
  • Interior with rounded corners to make cleaning easier
  • Door is lockable and reversible, self closing at 45° but stable at 90°
Lieferumfang: These refrigerator cabinets are delivered with adjustable feet (150 to 180 mm).
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
SERVICELEISTUNGEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge