Columns, ChiroSil®
Lieferant: ES INDUSTRIES
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
799103 799003 799102
ESIN799103EA 0 EUR InStock
ESIN799103 ESIN799003 ESIN799102
Columns, ChiroSil®
Chromatographiesäulen
These columns are used for the separation of amino acids and compounds containing primary amines in HPLC.
  • Exhibits excellent durability due to covalent bonding
  • Flow rate:1,0 ml/min
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge