SPE columns, PCB analysis, CHROMABOND® SA/SiOH
Lieferant: MACHEREY-NAGEL
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
730132 730132.250
MANA730132EA 277 EUR InStock
MANA730132 MANA730132.250
SPE columns, PCB analysis, CHROMABOND® SA/SiOH
Chromatographiesäulen
CHROMABOND® SA/SiOH is a combination phase for SPE analysis of PCBs.
  • SA cation exchanger for removal of ionic compounds
  • SiOH for removal of polar compounds
Application: PCB extraction from waste oil (hexane extract).
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge