Safety shoes, lace-up, Bacou Original Acanti
Lieferant: Honeywell Safety Products
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
DALL6246108/48EA 99.6 EUR InStock
DALL6246108/48 DALL6246108/47 DALL6246108/39 DALL6246108/49 DALL6246108/44 DALL6246108/43 DALL6246108/46 DALL6246108/45 DALL6246108/40 DALL6246108/42 DALL6246108/41
Safety shoes, lace-up, Bacou Original Acanti
Schuhe und Stiefel
Robust safety shoe made from perforated suede leather, padded ankle collar and tongue, lateral bellows and dual-density PU outsole PU 2D resistant to abrasion, fuel oil and grease. High slip resistance due to well defined cleat design, wide, deep and self-cleaning. Resistant to acids and alkalis: limited to low concentrations.
  • Antibacterial lining with 3D structure
  • Removeable, dual density PU foam insole
  • Shock absorber heel
  • Light, metal-free injected polymer Spacium™ toecap (200 J)
The shoe is slip resistant, and is recommended for people working in administration, industrial cleaning, logistics and maintenance.
Colour: Grey
Zertifizierungen: EN ISO 20345:2007; S1P, HI, CI, SRC
Order Now
PRODUKT
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge