Chelex® 100, sodium form
Lieferant: SIGMAALDRICH
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
C7901-25G C7901-100G C7901-500G C7901-50G
SIALC7901-25GEA 127 EUR InStock
SIALC7901-25G SIALC7901-100G SIALC7901-500G SIALC7901-50G
Chelex® 100, sodium form
Protein-Aufbereitungs Protein Purification Resins
Chelex® 100 Chelating Resin is a weak cation exchange resin that has high selectivity, approximately 5,000:1, for copper, iron, and other heavy metal ions over monovalent cations. Chelex 100 Resin is made of a styrene divinylbenzene copolymer containing paired iminodiacetate ions, which act as chelating groups in binding polyvalent metal ions.
There are three types of Chelex® 100 Chelating Resin available; analytical, biotechnology grade, and molecular biology grade. Analytical grade Chelex® 100 Resin is sized and purified for reproducibility in analytic techniques. Biotechnology grade Chelex® 100 Resin is the analytical grade form certified to contain fewer than 100 microorganisms per gram of resin.
Molecular biology grade Chelex® 100 Resin can be used for fast, easy DNA purification from many sample types, such as forensic, blood, mammalian cells, and bacteria, including samples with high levels of salts. The resin avidly binds divalent cations such as Mg2+, inactivating DNases and other enzymes as well as binding other compounds that can interfere with enzyme-based applications such as PCR and ligation.
The resin has a strong affinity for transition metals, even in highly concentrated salt solutions.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge