Trident direct guard cartridge system
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
25084 25082 25083 25087 25088
REST25084EA 0 EUR InStock
REST25084 REST25082 REST25083 REST25087 REST25088
Trident direct guard cartridge system
Chromatographie-Zubehör Chromatographie-Ersatzteile
Trident direct system can be used to select the right level of protection for analysis. It provides the protection against particulate matter and moderate protection against irreversibly adsorbed compounds.
  • Easy to use
  • Low dead volume
  • Leak-free cartridge
The system is available in configurations to match different protection levels such as in-line filter, in-line filter with holder for 10 mm guard cartridge. The guard cartridges are available in 2,1 and 4,0 mm ID and are interchangeable within the appropriate length holder.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge