Unleaded gasolines (ASTM Class 2 Accelerant)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
30096 30097 30098 30099 30436
REST30096EA 71.5 EUR InStock
REST30096 REST30097 REST30098 REST30099 REST30436
Unleaded gasolines (ASTM Class 2 Accelerant)
Standards Mineralölindustrie-Standards
The weathered materials indicate the percent weight loss from the original material. Prepared from a single source (one refinery) product.
Zertifizierungen: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge