8270 Matrix Spike Mix (76 components)
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
31687 31687.15 33073
REST31687EA 279.5 EUR InStock
REST31687 REST31687.15 REST33073
8270 Matrix Spike Mix (76 components)
Standards Organische Standards
The analysis of semivolatile organic pollutants in solid waste, soil, water, and air matrices using gas chromatography - mass spectrometry (GC - MS).
1,2-diphenylhydrazine decomposes to azobenzene (mix component) in the injector. N-nitrosodiphenylamine decomposes to diphenylamine (mix component) in the injector.
Zertifizierungen: EPA standards.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge