Kerosene composite standards
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
31094 31256 31257 31094-3 31094-5 31256-3 31256-5 31257-3 31257-5
REST31094EA 50 EUR InStock
REST31094 REST31256 REST31257 REST31094-3 REST31094-5 REST31256-3 REST31256-5 REST31257-3 REST31257-5
Kerosene composite standards
Standards Mineralölindustrie-Standards
Kerosene composite standards are comprised of three separate sources blended in equal portions. It may exhibit lot-to-lot variation.
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge