Ferrules, GRAPHPACK®
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
22223 22224
REST22223EA 0 EUR InStock
REST22223 REST22224
Ferrules, GRAPHPACK®
Fittings und Verbinder
GRAPHPACK® technology provides a complete system that quickly and reliably makes a leak-tight, low-dead-volume connection.
  • Ideal seal for Gerstel CIS 3 and CIS 4 PTV Inlets and also for GC applications
  • Central component is a metal-jacketed graphite ferrule
  • Fit columns with internal diameters ranging from 0,25 to 0,53 mm
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge