Trident in-line guard cartridge holders for HPLC columns
Lieferant: Restek
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
25021 25040
REST25021EA 85 EUR InStock
REST25021 REST25040
Trident in-line guard cartridge holders for HPLC columns
Vorsäulenhalter
A Trident in-line guard cartridge holder can be used with almost any HPLC column by connecting it with a short piece of 1/16" tubing, appropriate nuts and ferrules, or finger-tight fittings.
  • Suitable for 10 mm guard cartridges
Prefilter provides added protection against the particles that can shorten the lifetime of the guard cartridge.
Bestellinformation: Purchase with or without a prefilter, which provides added protection against the particles that can shorten the lifetime of the guard cartridge.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge