Programmable controllers
Lieferant: CARBOLITE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
02-3216P1 02-3216P5
CARB02-3216P1EA 0 EUR InStock
CARB02-3216P1 CARB02-3216P5
Programmable controllers
Regler Temperaturregler
These controllers for programmable temperature can use up to eight segments, each segment comprising a ramp followed by a dwell. The dwell may be set to zero time.
  • Multiple line numeric and alphamumeric display
  • Navigation via four push button controls
  • Simultaneous display of set-point and actual temperatures
  • Panel mounted device with a clear LED display
  • PID control technology
Alarm signal for high-low deviation, sensor break, event or element failure.
W×H: 48×48 mm
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge