Revised GPC calibration mix, semi volatile compounds
Lieferant: RESTEK
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
Sortieren nach:
32041 32042
REST32041EA 48.5 EUR InStock
REST32041 REST32042
Revised GPC calibration mix, semi volatile compounds
Standards Umweltanalyse-Standards
Revised GPC calibration mix is a qualitative mixture and it is useful for determining GPC dump/collect times. The compounds are dissolved in methylene chloride at various concentrations.
It is made up of five components: 5 mg/ml of Bis(2-ethylhexyl)phthalate, 250 mg/ml of corn oil, 1,0 mg/ml of methoxychlor, 0,2 mg/ml of perylene, 0,8 mg/ml of sulphur.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge